Learn More
Medchemexpress LLC Ralaniten triacetate | 1637573-04-6 | 98.8% | 521.00 | C27H33ClO8 | 50MG

Supplier: Medchemexpress LLC HY123875A50MG
Ralaniten triacetate (EPI-506) is a research-stage prodrug of ralaniten that inhibits the androgen receptor N-terminal domain (NTD). It is intended for preclinical biochemical and cellular research into AR signaling and prostate cancer models.
- CAS number: 1637573-04-6.
- Purity: 98.8% (reported by supplier).
- Molecular formula: C27H33ClO8.
- Molecular weight: 521.00 Da.
- Physical state: colorless to light yellow liquid; density ≈1.188 g/cm3.
- Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Pack sizes: available in milligram quantities for research use (including 50 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.