Learn More
Medchemexpress LLC Ralaniten triacetate | 1637573-04-6 | 98.8% | 521.00 g/mol | C27H33ClO8 | 100MG
SDP
Supplier:  Medchemexpress LLC HY123875A100MG
Encompass_Preferred
Ralaniten triacetate is the triacetate prodrug of ralaniten (EPI-506), an orally active inhibitor targeting the N-terminal domain (NTD) of the androgen receptor (AR). It is used in research to study AR signaling and has demonstrated activity against full-length AR and resistance-associated splice variants such as AR-v7. The compound is supplied in multiple solution concentrations and solid pack sizes for laboratory use.
  • Prodrug designed for improved oral bioavailability.
  • Targets the AR N-terminal domain to inhibit receptor activity.
  • Active against full-length and variant AR forms, including AR-v7.
  • Available in multiple solution concentrations and solid amounts for research use.
  • High reported purity suitable for biochemical and cellular studies.
Catalog No. 50-002-37147