Learn More
Selleck Chemical LLC Raloxifene, 25mg | CAS No. 84449-90-1 | Purity: 98.47%
SDP
Supplier:  Selleck Chemical LLC S578125MG
Encompass_Preferred
Raloxifene (Keoxifene, Pharoxifene, LY-139481, LY-156758, CCRIS-7129) is a second generation selective estrogen receptor modulator (SERM) used to prevent osteoporosis in postmenopausal women.
Catalog No. NC3662311