Learn More
Selleck Chemical LLC Raloxifene
SDP
Supplier:  Selleck Chemical LLC S57815MG
Encompass_Preferred
Raloxifene (Keoxifene Pharoxifene LY-139481 LY-156758 CCRIS-7129) is a second generation selective estrogen receptor modulator (SERM) used to prevent osteoporosis in postmenopausal women
Catalog No. 50-000-00027