Learn More
Enzo Life Sciences Raloxifene. HCl (50mg). CAS: 82640-04-8
SDP
Supplier:  Enzo Life Sciences BMLGR2430050
Encompass_Preferred
A selective estrogen receptor modulator (SERM). Displays anti-estrogen action along with anti-androgen action. In clinical use for prevention of postmenopausal osteoporosis. Reduces circulating levels of IL-6 and TNFα. Inhibits prostate carcinogenesis in transgenic rat model. May be useful for the prevention of hormone-related cancers. Purity: ≥98% (TLC). Solubility: Soluble in methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0947