Learn More
TARGETMOL CHEMICALS INC Raloxifene hydrochloride 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1549200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Raloxifene hydrochloride (LY156758 hydrochloride) is a second generation selective estrogen receptor modulator (SERM) used to prevent osteoporosis in postmenopausal women. It has estrogen agonist effects on bone and cholesterol metabolism but behaves as a complete estrogen antagonist on mammary gland and uterine tissue. Purity 99.81%
Catalog No. 50-287-8260