Learn More
Medchemexpress LLC Raloxifene hydrochloride | 82640-04-8 | MFCD01938233 | 99.8% | 510.0 g/mol | C28H28ClNO4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13738AR5MG
Encompass_Preferred
Raloxifene hydrochloride is the hydrochloride salt of raloxifene, a selective estrogen receptor modulator. This analytical standard is provided as a high-purity solid for research and analytical use, and is commonly supplied in small vial sizes (including 5 mg) for reference, QC, and method development.
  • High purity (~99.8%).
  • Suitable for HPLC and method validation.
  • Available in small vial sizes, including 5 mg.
  • Certificate of analysis and safety data sheet available.
  • Off-white to pale-yellow solid appearance.
Catalog No. 50-002-22808