Learn More
Cayman Chemical RaloxIfenhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2718910 MG
Encompass_Preferred
An analytical reference standard categorized as an SERM; has been detected as an AAF during anti-doping testing; intended for use in analytical forensic applications
Catalog No. 50-293-8547