Learn More
Medchemexpress LLC HY-10353 5mg Medchemexpress, Raltegravir CAS:518048-05-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103535MG
Encompass_Preferred
Medchemexpress, HY-10353 5mg Raltegravir CAS:518048-05-0 Raltegravir is a potent integrase (IN) inhibitor, used to treat HIV infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8124