Learn More
Selleck Chemical LLC Raltegravir (MK-0518) 10mM/1mL 518048-05-0
SDP
Supplier:  Selleck Chemical LLC S200510MM/1ML
Encompass_Preferred
Raltegravir (MK-0518) is a potent integrase (IN) inhibitor for WT and S217Q PFV IN with IC50 of 90 nM and 40 nM in cell-free assays, respectively. It shows greater than 1000-fold selectivity for HIV-1 IN over several related Mg2+-dependent enzyme such as HCV polymerase, HIV reverse transcriptase, HIV RNaseH and human ?-, ?-, ?-polymerases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1867