Learn More
Medchemexpress LLC Raltegravir potassium | 871038-72-1 | MFCD12031642 | 99.8% | 482.5 g/mol | C20H20FKN6O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10353A50MG
Encompass_Preferred
Raltegravir potassium is the potassium salt of raltegravir (MK-0518), a potent HIV-1 integrase strand transfer inhibitor supplied for laboratory research use only. It is provided in small pack sizes and is intended for in vitro research applications, not for human or veterinary use.
  • Potent integrase inhibitor used in HIV research.
  • CAS number 871038-72-1.
  • High purity (99.8%).
  • Available in small research pack sizes, with options for solid or DMSO solution formats.
  • Intended for research use only; not for clinical use.
Catalog No. 50-002-48199