Learn More
Medchemexpress LLC Raltegravir potassium | 871038-72-1 | MFCD12031642 | 99.8% | 482.51 g/mol | C20H20FKN6O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10353A100MG
Encompass_Preferred
Raltegravir potassium is the potassium salt form of raltegravir, a potent HIV integrase strand transfer inhibitor supplied as a research-grade solid for laboratory and analytical applications. The product is intended for research use only and includes storage recommendations and quality documentation.
  • Potent integrase strand transfer inhibitor used in HIV research.
  • Potassium salt form for improved solubility in aqueous media.
  • High chemical purity (approximately 99.8%).
  • Solid form with recommended storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Available in small pack sizes suitable for assay development and analytical work.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-48476