Learn More
Selleck Chemical LLC Raltitrexed 100mg 112887-68-0 ZD-1694
SDP
Supplier:  Selleck Chemical LLC S1192100MG
Encompass_Preferred
Raltitrexed (ZD-1694) is a thymidylate synthase inhibitor with an IC50 of 9 nM for the inhibition of L1210 cell growth. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-781-8