Learn More
Selleck Chemical LLC Ramelteon S1259-2mg
SDP
Supplier:  Selleck Chemical LLC S12592MG
Encompass_Preferred
Ramelteon (TAK-375) is a novel melatonin receptor agonist for human MT1 and MT2 receptors and chick forebrain melatonin receptors with Ki of 14 pM 112 pM and 23 1 pM respectively
Catalog No. 50-310-8343