Learn More
TARGETMOL CHEMICALS INC RAMELTEON 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T146325MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ramelteon (TAK-375) is a Melatonin Receptor Agonist. The mechanism of action of ramelteon is as a Melatonin Receptor Agonist. purity: 99%
Catalog No. 50-257-7380