Learn More
Medchemexpress LLC Ramelteon impurity 10 | 196597-17-8 | 98.7% | 259.35 | C16H21NO2 | 10 MG

Supplier: Medchemexpress LLC HY7525610MG
Ramelteon impurity 10 is an analytical reference impurity of ramelteon used for impurity profiling, method development, and quality control. Supplied for research use only with an accompanying certificate of analysis and safety data sheet; the COA reports purity by HPLC at 98.66%.
- Analytical reference for impurity profiling
- Suitable for HPLC and LC-MS method development
- CAS number 196597-17-8
- Molecular formula C16H21NO2
- Molecular weight 259.35 g/mol
- Certified purity 98.66% by HPLC (COA provided)
- Intended for research use only; not for human use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.