Learn More
Selleck Chemical LLC Ramipril S1793-50mg
SDP
Supplier:  Selleck Chemical LLC S179350MG
Encompass_Preferred
Ramipril(HOE-498) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 5 nM
Catalog No. 50-311-0764