Learn More
Apexbio Technology LLC Ramipril 87333-19-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22085.1
Encompass_Preferred
Ramipril (CAS 87333-19-5) is a small-molecule inhibitor of angiotensin-converting enzyme (ACE) a key enzyme in the renin-angiotensin system responsible for the conversion of angiotensin I to angiotensin II By inhibiting ACE activity with an IC50 of 5 nM ramipril effectively reduces the formation of angiotensin II a potent vasoconstrictor involved in blood pressure regulation and cardiovascular remodeling Ramipril is widely utilized in biomedical research to investigate mechanisms of hypertension cardiovascular diseases and the pharmacological modulation of the renin-angiotensin system
Catalog No. 50-000-89356