Learn More
Selleck Chemical LLC Ramulus Mori Extract E3603-5MG
SDP
Supplier:  Selleck Chemical LLC E36035MG
Encompass_Preferred
Ramulus Mori Extract is extracted from Ramulus mori (Mulberry twigs) the dry branch of Morus alba L Ramulus Mori is a traditional medicine widely used for diabetes treatment The major active components of Ramulus Mori are mulberroside A oxyresveratrol resveratrol 4-hydroxycinnamic acid 7-hydroxycoumarin and morin and the components have been demonstrated to possess anti-cancer anti-hypertensive anti-diabetic and anti-inflammatory properties
Catalog No. 50-000-40079