Learn More
Medchemexpress LLC Randialic acid B | 14021-14-8 | 1 MG

Supplier: Medchemexpress LLC HYN81521MG
Randialic acid B is a triterpenoid compound that functions as a formyl peptide receptor 1 (FPR1) antagonist. It selectively inhibits reactive oxygen species production, elastase release, and CD11b expression in human neutrophils. In vivo studies demonstrate its ability to significantly attenuate psoriasis-like inflammation.
- Purity of 99.94%
- Functions as a formyl peptide receptor 1 (FPR1) antagonist
- Selectively inhibits reactive oxygen species production, elastase release, and CD11b expression in human neutrophils
- Attenuates psoriasis-like inflammation in vivo
- Provides cell-protective effects against H2O2-induced H9c2 cardiomyocyte injury
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.