Learn More
Medchemexpress LLC Ranibizumab anti-VE 25mg | 25mg
SDP
Supplier:  Medchemexpress LLC HYP9951A25MG
Encompass_Preferred
Ranibizumab (RG-6321) (anti-VEGF) is a humanized anti-VEGF monoclonal antibody fragment and can recognize all VEGF-A isoforms (VEGF110 VEGF121 and VEGF165)[1] Ranibizumab (anti-VEGF) slows vision loss in vivo and is used for wet age-related macular degeneration (AMD) research[1]
Catalog No. 50-003-22823