Learn More
Selleck Chemical LLC Ranitidine
SDP
Supplier:  Selleck Chemical LLC S566225MG
Encompass_Preferred
Ranitidine(AH-19065) is an H2 histamine receptor antagonist that works by blocking histamine and thus decreasing the amount of acid released by cells of the stomach
Catalog No. 50-000-03700