Learn More
TARGETMOL CHEMICALS INC Ranitidine 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T361050MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Ranitidine (HSDB 3925) is a non-imidazole blocker of those histamine receptors that mediate gastric secretion (H2 receptors). It is used to treat gastrointestinal ulcers. Purity 99.1%
Catalog No. 50-287-6462