Learn More
Medchemexpress LLC Ranitidine | 66357-35-5 | ≥99.0% | 314.40 g/mol | C13H22N4O3S | 10 MG

Supplier: Medchemexpress LLC HYB069310MG
Ranitidine is a histamine H2-receptor antagonist used as a research reagent to inhibit gastric acid secretion and to study H2 receptor pharmacology. Supplied in solid and DMSO solution formats, it is intended for laboratory research use only.
- High purity (≥99.0%).
- Molecular weight 314.40 g/mol.
- Chemical formula C13H22N4O3S.
- Available in small solid quantities and DMSO solution preparations.
- Suitable for receptor pharmacology and gastric secretion studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.