Learn More
Medchemexpress LLC Ranitidine | 66357-35-5 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0693100MG
Encompass_Preferred
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It has been shown to antagonize histamine-induced responses in isolated rat tissues and inhibit breast tumor development and spread in mice.
  • Potent, selective, and orally active histamine H2-receptor antagonist
  • Inhibits gastric secretion
  • Antagonizes histamine-induced increases in guinea-pig isolated rat atrium
  • Antagonizes histamine-induced relaxations of the rat isolated uterine horn
  • Inhibits breast tumor development and spread in mice
  • Reduces CD11b+Ly6Chi cells in spleen and bone marrow
  • Inhibits lung metastasis in breast cancer models
  • Inhibits primary tumor growth in breast cancer models
  • Increases latency of breast tumorigenesis and reduces tumor numbers
Catalog No. 50-004-08916