Learn More
Medchemexpress LLC Ranitidine | 66357-35-5 | 99.9% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0693
Encompass_Preferred
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It antagonizes histamine-induced increases in guinea-pig isolated rat atrium and histamine-induced relaxations of the rat isolated uterine horn, with pA2 values of 7.2 and 6.95, respectively. It also inhibits breast tumor development and spread in mice.
  • Inhibits gastric secretion
  • Antagonizes histamine-induced increases in guinea-pig isolated rat atrium
  • Antagonizes histamine-induced relaxations of the rat isolated uterine horn
  • Inhibits breast tumor development and spread in mice
  • Reduces CD11b+Ly6Chi cells in spleen and bone marrow
  • Inhibits lung metastasis
  • Inhibits primary tumor growth
  • Increases the latency of breast tumorigenesis and reduces tumor count
Catalog No. 50-004-07435