Learn More
Apexbio Technology LLC Ranitidine 66357-59-3 100mg
SDP
Supplier:  Apexbio Technology LLC B1564100
Encompass_Preferred
Ranitidine (CAS 66357-59-3) is a small-molecule antagonist targeting histamine H2 receptors It is designed to competitively inhibit histamine binding to H2 receptors on gastric parietal cells thereby reducing gastric acid secretion Ranitidine exerts its biological activity primarily through blockade of histamine H2 receptors In experimental pharmacological assays ranitidine demonstrates antagonist activity with an IC50 value of approximately 3 3 1 4 M Based on these pharmacological properties ranitidine holds research potential in studies of gastric acid-related physiology gastrointestinal ulcer models and pharmacological interventions involving histaminergic pathways
Catalog No. 50-000-90575