Learn More
Medchemexpress LLC Ranitidine-d6 hydrochloride | 1185238-09-8 | 97.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0281AS1MG
Encompass_Preferred
Ranitidine-d6 hydrochloride is the deuterium labeled Ranitidine hydrochloride (HY-B0281A). Ranitidine hydrochloride is a potent, selective and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It antagonizes Histamine (HY-B1204)-induced increases of the guinea-pig isolated rat atrium and Histamine-induced relaxations of the rat isolated uterine horn, with pA2 values of 7.2 and 6.95, respectively. Ranitidine hydrochloride also inhibits breast tumor development and spread in mice.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Target: H2 receptor.
Catalog No. 50-004-08752