Learn More
Selleck Chemical LLC Ranitidine Hydrochloride S1801-50mg
SDP
Supplier:  Selleck Chemical LLC S180150MG
Encompass_Preferred
Ranitidine (AH19065) is a histamine H2-receptor antagonist used to treat stomach or intestinal ulcers
Catalog No. 50-311-0576