Learn More
Medchemexpress LLC Ranitidine S-oxide | 73851-70-4 | 99.0% | 330.41 | 10 MG

Supplier: Medchemexpress LLC HYW01124510MG
Ranitidine S-oxide is a metabolite of Ranitidine, a potent, selective, and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM, which inhibits gastric secretion.
- Metabolite of Ranitidine.
- Acts as a histamine H2-receptor antagonist.
- Inhibits gastric secretion.
- Molecular weight is 330.41.
- Purity of 99.03%.
- Appears as a yellow to brown solid.
- Soluble in water at 125 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.