Learn More
Medchemexpress LLC Ranitidine S-oxide | 73851-70-4 | 99.0% | 330.41 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0112455MG
Encompass_Preferred
Ranitidine S-oxide is a metabolite of Ranitidine (HY-B0693). Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM that inhibits gastric secretion.
  • Metabolite of Ranitidine.
  • Potent, selective, and orally active histamine H2-receptor antagonist.
  • Inhibits gastric secretion.
  • IC50 of 3.3 μM.
Catalog No. 50-004-53621