Learn More
Invivogen RANKL Reporter HEK 293 Cells
SDP
Supplier:  Invivogen HKBRANKL
Encompass
3-7 x 10e6 cells. Human & Mouse RANKL Reporter Cells. HEK-Blue RANKL cells were engineered from the human embryonic kidney HEK293 cell line to detect bioactive human and murine RANKL by monitoring the activation of the NF-κB and AP-1 pathways.
Catalog No. NC2741881