Learn More
Enzo Life Sciences RANKL (total), soluble (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850019KI01
Encompass_Preferred
The RANKL (total), soluble (human) ELISA kit is a complete assay for the quantitative determination of free and OPG-bound human soluble RANKL levels. This kit is to be used with serum, plasma and cell culture supernatant from human origin. Alternative name: ODF, OPGL, TRANCE, TNFSF 11, CD254. Assay Time: Overnight + 3 hours. Applications: ELISA . Species reactivity: Human .
Catalog No. 50-201-0092