Learn More
Selleck Chemical LLC Ranolazine 2HCl 100mg 95635-56-6 RS-43285
SDP
Supplier:  Selleck Chemical LLC S1425100MG
Encompass_Preferred
Ranolazine 2HCl (RS-43285) is a calcium uptake inhibitor via the sodium/calcium channal, used to treat chronic angina. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-631-5