Learn More
Medchemexpress LLC Ranolazine | 95635-55-5 | 99.8% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0280200MG
Encompass_Preferred
Ranolazine (CVT 303) is an anti-angina drug that inhibits the late phase of inward sodium current (INa and IKr) and acts as an antianginal agent. It is also a partial fatty acid oxidation (FAO) inhibitor. This product is for research use only and not sold to patients.
  • Inhibits late phase of inward sodium current (INa and IKr)
  • Acts as an antianginal agent
  • Partial fatty acid oxidation (FAO) inhibitor
  • Does not affect heart rate or blood pressure
Catalog No. 50-004-05836