Learn More
Abcam Rapamycin, mTORC1 comple x inhibitor, 1MG
Supplier:  Abcam AB1202241MG
Encompass_Preferred
MW 914.2 Da, Purity >98%. Immunosuppressant and antifungal agent . Complexes with FKBP-12 and inhibits mTOR (mammalian target of rapamycin). In complex with its cellular receptor, the FK506-binding protein (FKBP12), rapamycin binds and inhibits the function of the mammalian target of rapamycin (mTOR).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4646