Learn More
Selleck Chemical LLC Rapamycin (Sirolimus) 25mg 53123-88-9 AY 22989NSC-2260804
SDP
Supplier:  Selleck Chemical LLC S103925MG
Encompass_Preferred
Rapamycin (Sirolimus, AY 22989, NSC-2260804) is a specific mTOR inhibitor with IC50 of ~0.1 nM HEK293 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0900090