Learn More
Medchemexpress LLC Rapamycin (standard) | 53123-88-9 | MFCD00867594 | 99.0% | 914.2 g/mol | C51H79NO13 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10219R25MG
Encompass_Preferred
Rapamycin (sirolimus) analytical standard supplied as a purified reference material for research and analytical applications. It is a macrocyclic lactone mTOR inhibitor that binds FKBP12 and acts as an allosteric inhibitor of mTORC1; reported IC50 in HEK293 cells is 0.1 nM. Intended for assay, calibration, method development, and analytical quality control.
  • Analytical standard suitable for assay and calibration.
  • Purity 98.98% (manufacturer reported).
  • Available as a 25 mg solid powder for laboratory use.
  • Suitable for mTOR pathway studies and method validation.
Catalog No. 50-002-46348