Learn More
Selleck Chemical LLC Rapastinel
SDP
Supplier:  Selleck Chemical LLC S69075MG
Encompass_Preferred
Rapastinel (GLYX-13 BV-102 TPPT-amide) is a partial agonist of N-methyl-D-aspartate receptor (NMDAR) that targeting a glycine site Rapastinel has long-lasting antidepressant effects
Catalog No. 50-000-00697