Learn More
Key Scientific Products Rapid Carbapenemase Screen (Kit), 25 Tablets Equivalent to 50 Tests
SDP
Supplier:  Key Scientific Products RO98024
Encompass_Preferred
The Rapid CARB kit is a stabilized CARBA-NP test that detects the carbapenemase resistance mechanism in Enterobacteriaceae and Pseudomonas aeruginosa.
Catalog No. NC0870932