Learn More
Sigma Aldrich Fine Chemicals Biosciences Rapid DNA Dephos Ligatio
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4898117001
Encompass
The Rapid DNA Dephos Ligation Kit enables fast and efficient dephosphorylation and ligation of sticky- or blunt-end DNA fragments.
Catalog No. 50-290-5533