Learn More
PROTEIOS TECHNOLOGY INC RapidPro™ Gravity Kit
Supplier:  PROTEIOS TECHNOLOGY INC SQ3311355
Encompass
The RapidPro™ Gravity Kit uses Car9-tag affinity for fast, silica-based protein purification, achieving >98% purity in 30-45 min with no specialized equipment or hazardous chemicals.
Catalog No. NC3591620