Learn More
Waters Corp Waters Glycan Standard, Quantitative, 100 pmol, HILIC Chromatography, GU Value 4
Supplier:  Waters Corp 186008791
Encompass_Preferred
The Quantitative Test Standard is designed to quantify absolute amounts of labeled analytes using fluorescence response and to assess the yields and proficiency of sample preparations. Each vial contains a lyophilized, highly purified synthetic peptide derivatized with a single label. It is designed to elute via HILIC chromatography with a GU (glucose unit) value of approximately 4 and is QC-tested to contain 100 pmol per vial.
  • Intended for quantifying absolute amounts of labeled analytes.
  • Assesses yields and proficiency of sample preparations.
  • Contains a lyophilized, highly purified synthetic peptide.
  • Derivatized with a single label.
  • Designed to elute via HILIC chromatography.
  • QC-tested to ensure 100 pmol per vial.
  • Has a GU (glucose unit) value of approximately 4.
Catalog No. 50-146-1956