Learn More
Medchemexpress LLC RAPTA-C (Ru(η6-p-cymene)Cl2(pta)) | 372948-28-2 | 99.3% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1303261MG
Encompass_Preferred
RAPTA-C acts as an anti-cancer and anti-angiogenic agent. It exhibits anti-metastatic, anti-angiogenic, and anti-tumoral activities through protein and histone-deoxyribonucleic acid alterations.
  • Inhibits cell growth by triggering G(2)/M phase arrest in cancer cells
  • Enhances p53 levels
  • Triggers the mitochondrial apoptotic pathway
  • Reduces tumor growth with inhibition of angiogenesis in an ovarian carcinoma model
Catalog No. 50-003-39101