Learn More
Medchemexpress LLC HY-121320 5mg , Raptinal CAS:1176-09-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1213205MG
Encompass_Preferred
HY-121320 5mg Raptinal CAS: 1176-09-6 Raptinal, a agent that directly activates caspase-3, initiates intrinsic pathway caspase-dependent apoptosis. Raptinal is able to rapidly induce cancer cell death by directly activating the effector caspase-3, bypassing the activation of initiator caspase-8 and caspase-9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9983