Learn More
Rarecyte RareCyte AccuCyte Blood Collection Tube, Sterile, 100-Pack
Supplier:  Rarecyte 241208100
Encompass
A pack of hundred sterile AccuCyte Blood Collection Tubes (BCTs). Contents in the tubes stabilize and preserve nucleated cells in whole blood.
Catalog No. NC2069161