Learn More
Rarecyte RareCyte CytePicker Chamber Slide, 4-Well, 10-Pack
Supplier:  Rarecyte 241068000
Encompass
A pack of ten Imaging chamber slides, for picking cells from solution. Each slide has two large sample wells, two small repick wells, and holders for three PCR tubes. Compatible with both, CyteFinder I and CyteFinder II instruments.
Catalog No. NC2069220