Learn More
ABclonal Technology RARS Rabbit mAb
SDP
Supplier:  ABclonal Technology A2633320UL
Encompass_Preferred
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution Arginyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family
Catalog No. 50-000-08860