Learn More
Medchemexpress LLC Rasagiline | 136236-51-6 | 99.9% | 171.24 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY14605A
Encompass_Preferred
Rasagiline is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor. It also acts as a click chemistry reagent, featuring an Alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Highly potent selective irreversible mitochondrial monoamine oxidase inhibitor
  • Inhibits MAO B activity with IC50 of 4.43 nM
  • Inhibits MAO A activity with IC50 of 412 nM
  • Functions as a click chemistry reagent
  • Capable of copper-catalyzed azide-alkyne cycloaddition
  • Demonstrates neuroprotective properties in transgenic models
  • Increases cell proliferation rates in SH-SY5Y and 1242-MG cells
Catalog No. 50-003-63435