Learn More
Medchemexpress LLC Rasagiline-13C3 mesylate | 1391052-18-8 | 270.32 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14605S1MG
Encompass_Preferred
Rasagiline-13C3 mesylate is a 13C-labeled version of Rasagiline (mesylate). It acts as a highly potent, selective, and irreversible mitochondrial monoamine oxidase (MAO) inhibitor.
  • 13C-labeled version of Rasagiline (mesylate)
  • Highly potent MAO inhibitor
  • Selective MAO inhibitor
  • Irreversible mitochondrial MAO inhibitor
  • IC50 values of 4.43 nM for rat brain MAO B and 412 nM for MAO A activity
Catalog No. 50-003-59765